Assay Details
| Gene Symbol: camk2d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003602.2 | NP_001003602.1 | 15 - 16 | 1316 | 73 | |
| NM_001114708.2 | NP_001108180.1 | 16 - 17 | 1361 | 73 | ||
| XM_005166561.4 | XP_005166618.1 | 15 - 16 | 1352 | 73 | ||
| XM_009303481.3 | XP_009301756.1 | 17 - 18 | 1699 | 73 | ||
| XM_009303482.3 | XP_009301757.1 | 17 - 18 | 1700 | 73 | ||
| XM_009303484.3 | XP_009301759.1 | 16 - 17 | 1654 | 73 | ||
| XM_009303485.3 | XP_009301760.1 | 16 - 17 | 1397 | 73 | ||
| XM_021477468.1 | XP_021333143.1 | 17 - 18 | 1458 | 73 | ||
| GenBank mRNA | BC077143.1 | - | 15 - 16 | 1284 | 73 | |
| BC154189.1 | - | 16 - 17 | 1360 | 73 | ||
| EE714463.1 | - | 3 - 4 | 175 | 73 | ||
More Information
Set Membership: |
|

camk2d1
Genomic Map