Assay Details
| Gene Symbol: camk2d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003602.2 | NP_001003602.1 | 6 - 7 | 623 | 86 | |
| NM_001114708.2 | NP_001108180.1 | 6 - 7 | 623 | 86 | ||
| XM_009303481.3 | XP_009301756.1 | 6 - 7 | 661 | 86 | ||
| XM_009303484.3 | XP_009301759.1 | 6 - 7 | 661 | 86 | ||
| XM_021477468.1 | XP_021333143.1 | 6 - 7 | 660 | 86 | ||
| GenBank mRNA | BC077143.1 | - | 6 - 7 | 591 | 86 | |
| BC154189.1 | - | 6 - 7 | 622 | 86 | ||
| CN513429.1 | - | 6 - 7 | 585 | 86 | ||
More Information
Set Membership: |
|

camk2d1
Genomic Map