Assay Details
| Gene Symbol: camk2d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003602.2 | NP_001003602.1 | 8 - 9 | 809 | 63 | |
| NM_001114708.2 | NP_001108180.1 | 8 - 9 | 809 | 63 | ||
| XM_005166561.4 | XP_005166618.1 | 8 - 9 | 845 | 63 | ||
| XM_009303481.3 | XP_009301756.1 | 8 - 9 | 847 | 63 | ||
| XM_009303482.3 | XP_009301757.1 | 8 - 9 | 848 | 63 | ||
| XM_009303484.3 | XP_009301759.1 | 8 - 9 | 847 | 63 | ||
| XM_009303485.3 | XP_009301760.1 | 8 - 9 | 845 | 63 | ||
| XM_021477468.1 | XP_021333143.1 | 8 - 9 | 846 | 63 | ||
| GenBank mRNA | BC077143.1 | - | 8 - 9 | 777 | 63 | |
| BC154189.1 | - | 8 - 9 | 808 | 63 | ||
| Gene Symbol: camk2b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | CK687938.1 | - | 5 - 6 | 367 | 63 | |
More Information
Set Membership: |
|

camk2d1
Genomic Map