Assay Details
| Gene Symbol: camk2d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003602.2 | NP_001003602.1 | 10 - 11 | 1023 | 81 | |
| NM_001114708.2 | NP_001108180.1 | 10 - 11 | 1023 | 81 | ||
| XM_005166561.4 | XP_005166618.1 | 10 - 11 | 1059 | 81 | ||
| XM_009303481.3 | XP_009301756.1 | 10 - 11 | 1061 | 81 | ||
| XM_009303482.3 | XP_009301757.1 | 10 - 11 | 1062 | 81 | ||
| XM_009303484.3 | XP_009301759.1 | 10 - 11 | 1061 | 81 | ||
| XM_009303485.3 | XP_009301760.1 | 10 - 11 | 1059 | 81 | ||
| XM_021477468.1 | XP_021333143.1 | 10 - 11 | 1060 | 81 | ||
| GenBank mRNA | BC077143.1 | - | 10 - 11 | 991 | 81 | |
| EB951992.1 | - | 1 - 2 | 57 | 81 | ||
| Gene Symbol: camk2b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | CK687938.1 | - | 7 - 8 | 581 | 81 | |
More Information
Set Membership: |
|

camk2d1
Genomic Map