Assay Details
Gene Symbol: snrpf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003881.3 | NP_001003881.1 | 3 - 4 | 383 | 120 | |
GenBank mRNA | AY648838.1 | - | 3 - 4 | 388 | 120 | |
BC162363.1 | - | 3 - 4 | 385 | 120 | ||
BC162369.1 | - | 3 - 4 | 385 | 120 | ||
EH472304.1 | - | 1 - 2 | 257 | 120 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
