Assay Details
Gene Symbol: ak1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003993.1 | NP_001003993.1 | 5 - 6 | 494 | 89 | |
XM_005168682.4 | XP_005168739.1 | 4 - 5 | 391 | 89 | ||
XM_005168683.4 | XP_005168740.1 | 5 - 6 | 575 | 89 | ||
XM_009302056.3 | XP_009300331.1 | 5 - 6 | 491 | 89 | ||
XM_017356070.2 | XP_017211559.1 | 6 - 7 | 704 | 89 | ||
GenBank mRNA | BC080261.1 | - | 5 - 6 | 494 | 89 | |
BC163982.1 | - | 3 - 4 | 351 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
