Assay Details
Gene Symbol: msna | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004296.1 | NP_001004296.1 | 13 - 14 | 1722 | 67 | |
XM_009301281.3 | XP_009299556.1 | 13 - 14 | 1620 | 67 | ||
XM_009301282.3 | XP_009299557.1 | 9 - 10 | 1165 | 67 | ||
GenBank mRNA | BC081551.1 | - | 13 - 14 | 1722 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
