Assay Details
Gene Symbol: msna | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004296.1 | NP_001004296.1 | 4 - 5 | 460 | 71 | |
XM_009301281.3 | XP_009299556.1 | 4 - 5 | 358 | 71 | ||
GenBank mRNA | BC049479.1 | - | 4 - 5 | 524 | 71 | |
BC081551.1 | - | 4 - 5 | 460 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
