Assay Details
| Gene Symbol: ccnb1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004609.1 | NP_001004609.1 | 500 | 88 | ||
| GenBank mRNA | BC081485.1 | - | 500 | 88 | ||
| BC164004.1 | - | 442 | 88 | |||
More Information
Set Membership: |
|

ccnb1l
Genomic Map