Assay Details
Gene Symbol: pttg1ipb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005941.1 | NP_001005941.1 | 4 - 5 | 434 | 63 | |
GenBank mRNA | BC083469.1 | - | 4 - 5 | 434 | 63 | |
BC164885.1 | - | 4 - 5 | 388 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
