Assay Details
| Gene Symbol: eef2kmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006058.1 | NP_001006058.1 | 4 - 5 | 428 | 70 | |
| XM_009299451.2 | XP_009297726.1 | 4 - 5 | 617 | 70 | ||
| GenBank mRNA | BC083266.1 | - | 4 - 5 | 428 | 70 | |
More Information
Set Membership: |
|

eef2kmt
Genomic Map