Assay Details
| Gene Symbol: syngr1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006067.1 | NP_001006067.1 | 2 - 3 | 583 | 64 | |
| XM_005171578.4 | XP_005171635.1 | 2 - 3 | 613 | 64 | ||
| GenBank mRNA | BC083255.1 | - | 2 - 3 | 583 | 64 | |
| BC165917.1 | - | 2 - 3 | 364 | 64 | ||
More Information
Set Membership: |
|

syngr1a
Genomic Map