Assay Details
Gene Symbol: phf23a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013499.1 | NP_001013517.1 | 2 - 3 | 569 | 62 | |
XM_021476141.1 | XP_021331816.1 | 2 - 3 | 215 | 62 | ||
GenBank mRNA | BC091667.1 | - | 2 - 3 | 569 | 62 | |
BC153421.1 | - | 3 - 4 | 739 | 62 | ||
BC163957.1 | - | 2 - 3 | 92 | 62 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021476141.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
