Assay Details
| Gene Symbol: pop5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013548.1 | NP_001013566.1 | 4 - 5 | 428 | 75 | |
| GenBank mRNA | BC091467.1 | - | 4 - 5 | 428 | 75 | |
More Information
Set Membership: |
|

pop5
Genomic Map