Assay Details
| Gene Symbol: phactr3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020537.1 | NP_001018373.1 | 10 - 11 | 1449 | 64 | |
| XM_009305736.3 | XP_009304011.1 | 10 - 11 | 1472 | 64 | ||
| XM_021479616.1 | XP_021335291.1 | 10 - 11 | 1481 | 64 | ||
| XM_021479617.1 | XP_021335292.1 | 10 - 11 | 1459 | 64 | ||
| GenBank mRNA | BC095090.1 | - | 10 - 11 | 1449 | 64 | |
More Information
Set Membership: |
|

phactr3b
Genomic Map