Assay Details
| Gene Symbol: ctns | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020571.1 | NP_001018407.1 | 8 - 9 | 1020 | 71 | |
| GenBank mRNA | BC095197.1 | - | 8 - 9 | 1020 | 71 | |
More Information
Set Membership: |
|

ctns
Genomic Map