Assay Details
| Gene Symbol: polr1e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020704.1 | NP_001018540.1 | 11 - 12 | 1105 | 67 | |
| GenBank mRNA | BC095836.1 | - | 11 - 12 | 1105 | 67 | |
| BC153550.1 | - | 11 - 12 | 1078 | 67 | ||
| BC164721.1 | - | 11 - 12 | 1104 | 67 | ||
More Information
Set Membership: |
|

polr1e
Genomic Map