Assay Details
| Gene Symbol: mrpl44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030260.2 | NP_001025431.2 | 4 - 5 | 895 | 63 | |
| GenBank mRNA | BC097173.1 | - | 4 - 5 | 702 | 63 | |
| BC133990.1 | - | 4 - 5 | 833 | 63 | ||
More Information
Set Membership: |
|

mrpl44
Genomic Map