Assay Details
| Gene Symbol: crfb12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040353.1 | NP_001035443.1 | 7 - 8 | 1052 | 78 | |
| GenBank mRNA | BC098871.1 | - | 7 - 8 | 1054 | 78 | |
| BC115200.1 | - | 7 - 8 | 1052 | 78 | ||
More Information
Set Membership: |
|

crfb12
Genomic Map