Assay Details
| Gene Symbol: cnga2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044746.1 | NP_001038211.1 | 2 - 3 | 205 | 73 | |
| GenBank mRNA | DQ866815.1 | - | 2 - 3 | 205 | 73 | |
More Information
Set Membership: |
|

cnga2a
Genomic Map