Assay Details
Gene Symbol: cnga2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044746.1 | NP_001038211.1 | 4 - 5 | 493 | 104 | |
GenBank mRNA | DQ866815.1 | - | 4 - 5 | 493 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
