Assay Details
| Gene Symbol: vegfab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001328597.1 | NP_001315526.1 | 2 - 3 | 773 | 72 | |
| NM_001328598.1 | NP_001315527.1 | 2 - 3 | 773 | 72 | ||
| XM_005164518.4 | XP_005164575.1 | 2 - 3 | 836 | 72 | ||
| XM_009299968.3 | XP_009298243.1 | 2 - 3 | 834 | 72 | ||
| GenBank mRNA | BC117665.1 | - | 2 - 3 | 103 | 72 | |
| BC139619.1 | - | 2 - 3 | 738 | 72 | ||
| DQ026828.1 | - | 2 - 3 | 758 | 72 | ||
| DQ150576.1 | - | 2 - 3 | 108 | 72 | ||
More Information
Set Membership: |
|

vegfab
Genomic Map