Assay Details
| Gene Symbol: vegfab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001328597.1 | NP_001315526.1 | 7 - 8 | 1262 | 137 | |
| GenBank mRNA | BC117665.1 | - | 7 - 8 | 592 | 137 | |
| DQ026828.1 | - | 6 - 7 | 1190 | 137 | ||
More Information
Set Membership: |
|

vegfab
Genomic Map