Assay Details
| Gene Symbol: vegfab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001328597.1 | NP_001315526.1 | 8 - 9 | 1407 | 72 | |
| NM_001328598.1 | NP_001315527.1 | 6 - 7 | 1233 | 72 | ||
| XM_005164518.4 | XP_005164575.1 | 7 - 8 | 1362 | 72 | ||
| XM_009299968.3 | XP_009298243.1 | 6 - 7 | 1291 | 72 | ||
| GenBank mRNA | BC117665.1 | - | 8 - 9 | 737 | 72 | |
| DQ026828.1 | - | 7 - 8 | 1335 | 72 | ||
More Information
Set Membership: |
|

vegfab
Genomic Map