Assay Details
Gene Symbol: klhl38a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044885.1 | NP_001038350.1 | 1 - 2 | 1356 | 72 | |
XM_021468086.1 | XP_021323761.1 | 2 - 3 | 2167 | 72 | ||
GenBank mRNA | BC171663.1 | - | 1 - 2 | 1455 | 72 | |
BC171665.1 | - | 1 - 2 | 1455 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
