Assay Details
| Gene Symbol: klhl38a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044885.1 | NP_001038350.1 | 2 - 3 | 1458 | 71 | |
| XM_021468086.1 | XP_021323761.1 | 3 - 4 | 2269 | 71 | ||
| GenBank mRNA | BC171663.1 | - | 2 - 3 | 1557 | 71 | |
| BC171665.1 | - | 2 - 3 | 1557 | 71 | ||
More Information
Set Membership: |
|

klhl38a
Genomic Map