Assay Details
| Gene Symbol: ppfibp1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045067.1 | NP_001038532.1 | 4 - 5 | 494 | 103 | |
| XM_005164661.3 | XP_005164718.1 | 5 - 6 | 724 | 103 | ||
More Information
Set Membership: |
|

ppfibp1a
Genomic Map