Assay Details
| Gene Symbol: pcgf5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045235.1 | NP_001038700.1 | 5 - 6 | 619 | 78 | |
| XM_005156115.4 | XP_005156172.1 | 5 - 6 | 765 | 78 | ||
| XM_009306506.2 | XP_009304781.1 | 2 - 3 | 111 | 78 | ||
| GenBank mRNA | BC116619.1 | - | 5 - 6 | 619 | 78 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_009306506.2 transcripts.
Set Membership: |
|

pcgf5b
Genomic Map