Assay Details
Gene Symbol: znf740b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076631.1 | NP_001070099.1 | 4 - 5 | 520 | 78 | |
XM_005162390.4 | XP_005162447.1 | 4 - 5 | 615 | 78 | ||
XM_005162391.4 | XP_005162448.1 | 3 - 4 | 394 | 78 | ||
XM_021469848.1 | XP_021325523.1 | 4 - 5 | 498 | 78 | ||
GenBank mRNA | BC124428.1 | - | 4 - 5 | 520 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
