Assay Details
| Gene Symbol: nsrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077430.2 | NP_001070898.1 | 6 - 7 | 625 | 67 | |
| GenBank mRNA | BC076130.1 | - | 6 - 7 | 645 | 67 | |
| BC092757.1 | - | 6 - 7 | 626 | 67 | ||
| BC110090.1 | - | 6 - 7 | 637 | 67 | ||
| BC124124.1 | - | 6 - 7 | 630 | 67 | ||
More Information
Set Membership: |
|

nsrp1
Genomic Map