Assay Details
| Gene Symbol: ube2l3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082797.1 | NP_001076266.1 | 2 - 3 | 418 | 64 | |
| XM_005165039.4 | XP_005165096.1 | 2 - 3 | 184 | 64 | ||
| GenBank mRNA | BC052104.1 | - | 2 - 3 | 378 | 64 | |
| BC056724.1 | - | 108 | 64 | |||
More Information
Set Membership: |
|

ube2l3b
Genomic Map