Assay Details
Gene Symbol: kcnf1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082811.1 | NP_001076280.1 | 1 | 1149 | 91 | |
XM_021468440.1 | XP_021324115.1 | 2 | 1528 | 91 | ||
GenBank mRNA | BC163424.1 | - | 1 | 1181 | 91 | |
BC163435.1 | - | 1 | 1181 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
