Assay Details
| Gene Symbol: prkacaa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082840.1 | NP_001076309.1 | 1 - 2 | 613 | 105 | |
| GenBank mRNA | BC133159.1 | - | 1 - 2 | 613 | 105 | |
More Information
Set Membership: |
|

prkacaa
Genomic Map