Assay Details
| Gene Symbol: prkacaa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082840.1 | NP_001076309.1 | 5 - 6 | 992 | 90 | |
| XM_021470955.1 | XP_021326630.1 | 5 - 6 | 751 | 90 | ||
| GenBank mRNA | BC133159.1 | - | 5 - 6 | 992 | 90 | |
More Information
Set Membership: |
|

prkacaa
Genomic Map