Assay Details
| Gene Symbol: prkacaa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082840.1 | NP_001076309.1 | 7 - 8 | 1219 | 95 | |
| XM_021470955.1 | XP_021326630.1 | 7 - 8 | 978 | 95 | ||
| GenBank mRNA | BC133159.1 | - | 7 - 8 | 1219 | 95 | |
 Back To Top
 Back To TopMore Information
| Set Membership: |  Amplicon length between 86 and 100  Mammalian Gene Collection  Detect Genomic DNA  Danio rerio  Probe spans exons | 
Gene Ontology Categories:
 Back To Top
 Back To Top
 prkacaa
prkacaa Genomic Map
Genomic Map Back To Top
 Back To Top