Assay Details
Gene Symbol: rbpjb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083551.1 | NP_001077020.1 | 4 - 5 | 388 | 75 | |
GenBank mRNA | AY332618.1 | - | 4 - 5 | 388 | 75 | |
BC163294.1 | - | 4 - 5 | 365 | 75 | ||
BC163296.1 | - | 4 - 5 | 365 | 75 | ||
BC167436.1 | - | 4 - 5 | 393 | 75 | ||
DQ388902.1 | - | 4 - 5 | 390 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
