Assay Details
| Gene Symbol: rbpjb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083551.1 | NP_001077020.1 | 10 - 11 | 1210 | 103 | |
| GenBank mRNA | AY332618.1 | - | 10 - 11 | 1210 | 103 | |
| BC163294.1 | - | 10 - 11 | 1187 | 103 | ||
| BC163296.1 | - | 10 - 11 | 1187 | 103 | ||
| BC167436.1 | - | 10 - 11 | 1215 | 103 | ||
More Information
Set Membership: |
|

rbpjb
Genomic Map