Assay Details
| Gene Symbol: akap1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098179.2 | NP_001091649.2 | 2 - 3 | 1702 | 69 | |
| XM_017350962.2 | XP_017206451.1 | 2 - 3 | 1709 | 69 | ||
| GenBank mRNA | BC139889.1 | - | 3 - 4 | 1361 | 69 | |
More Information
Set Membership: |
|

akap1b
Genomic Map