Assay Details
| Gene Symbol: adcy2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099987.1 | NP_001093457.1 | 15 - 16 | 2090 | 67 | |
| XM_005170038.4 | XP_005170095.1 | 15 - 16 | 2284 | 67 | ||
| XM_021468083.1 | XP_021323758.1 | 11 - 12 | 1229 | 67 | ||
More Information
Set Membership: |
|

adcy2b
Genomic Map