Assay Details
Gene Symbol: adcy2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099987.1 | NP_001093457.1 | 13 - 14 | 1912 | 119 | |
XM_005170038.4 | XP_005170095.1 | 13 - 14 | 2106 | 119 | ||
XM_021468083.1 | XP_021323758.1 | 9 - 10 | 1051 | 119 | ||
XM_021468084.1 | XP_021323759.1 | 13 - 14 | 2128 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
