Assay Details
| Gene Symbol: panx1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100030.2 | NP_001093500.1 | 3 - 4 | 568 | 108 | |
| GenBank mRNA | EH579520.1 | - | 3 - 4 | 601 | 108 | |
More Information
Set Membership: |
|

panx1b
Genomic Map