Assay Details
| Gene Symbol: ryr1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102571.1 | NP_001096041.1 | 18 - 19 | 2163 | 76 | |
| XM_017351957.2 | XP_017207446.1 | 19 - 20 | 2392 | 76 | ||
| XM_017351958.2 | XP_017207447.1 | 19 - 20 | 2392 | 76 | ||
| XM_017351959.2 | XP_017207448.1 | 19 - 20 | 2392 | 76 | ||
| XM_017351960.2 | XP_017207449.1 | 19 - 20 | 2392 | 76 | ||
| XM_017351961.2 | XP_017207450.1 | 19 - 20 | 2392 | 76 | ||
| XM_017351962.2 | XP_017207451.1 | 18 - 19 | 2371 | 76 | ||
| XM_017351963.2 | XP_017207452.1 | 18 - 19 | 2371 | 76 | ||
| XM_021467693.1 | XP_021323368.1 | 19 - 20 | 2392 | 76 | ||
| XM_021467694.1 | XP_021323369.1 | 19 - 20 | 2392 | 76 | ||
| XM_021467695.1 | XP_021323370.1 | 18 - 19 | 2371 | 76 | ||
| XM_021467696.1 | XP_021323371.1 | 19 - 20 | 2392 | 76 | ||
| GenBank mRNA | AB247454.1 | - | 18 - 19 | 2163 | 76 | |
More Information
Set Membership: |
|

ryr1b
Genomic Map