Assay Details
| Gene Symbol: cept1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109717.1 | NP_001103187.1 | 4 - 5 | 842 | 66 | |
| XM_009295764.3 | XP_009294039.1 | 4 - 5 | 1054 | 66 | ||
| XM_021469371.1 | XP_021325046.1 | 4 - 5 | 873 | 66 | ||
| GenBank mRNA | BC151968.1 | - | 4 - 5 | 842 | 66 | |
More Information
Set Membership: |
|

cept1b
Genomic Map