Assay Details
| Gene Symbol: cept1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109717.1 | NP_001103187.1 | 5 - 6 | 979 | 98 | |
| GenBank mRNA | BC151968.1 | - | 5 - 6 | 979 | 98 | |
More Information
Set Membership: |
|

cept1b
Genomic Map