Assay Details
| Gene Symbol: dlc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130944.1 | NP_571019.1 | 7 - 8 | 2126 | 88 | |
| GenBank mRNA | AF146429.1 | - | 7 - 8 | 2126 | 88 | |
| BC162095.1 | - | 7 - 8 | 2055 | 88 | ||
| BC162100.1 | - | 7 - 8 | 2055 | 88 | ||
| CU458991.1 | - | 7 - 8 | 2126 | 88 | ||
More Information
Set Membership: |
|

dlc
Genomic Map