Assay Details
| Gene Symbol: alcama | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131000.1 | NP_571075.1 | 13 - 14 | 1643 | 69 | |
| GenBank mRNA | BC050482.1 | - | 13 - 14 | 1579 | 69 | |
| BC071276.1 | - | 13 - 14 | 1644 | 69 | ||
| CU458962.1 | - | 13 - 14 | 1590 | 69 | ||
| L25273.1 | - | 13 - 14 | 1643 | 69 | ||
More Information
Set Membership: |
|

alcama
Genomic Map