Assay Details
| Gene Symbol: alcama | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131000.1 | NP_571075.1 | 8 - 9 | 1136 | 87 | |
| XM_005172635.4 | XP_005172692.1 | 8 - 9 | 1162 | 87 | ||
| GenBank mRNA | BC050482.1 | - | 8 - 9 | 1072 | 87 | |
| BC071276.1 | - | 8 - 9 | 1137 | 87 | ||
| CU458879.1 | - | 8 - 9 | 977 | 87 | ||
| CU458962.1 | - | 8 - 9 | 1083 | 87 | ||
| L25273.1 | - | 8 - 9 | 1136 | 87 | ||
More Information
Set Membership: |
|

alcama
Genomic Map