Assay Details
| Gene Symbol: spon2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131007.1 | NP_571082.1 | 2 - 3 | 267 | 93 | |
| GenBank mRNA | AB006084.1 | - | 2 - 3 | 267 | 93 | |
| KM655591.1 | - | 1 - 2 | 220 | 93 | ||
More Information
Set Membership: |
|

spon2a
Genomic Map