Assay Details
| Gene Symbol: cahz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131110.1 | NP_571185.1 | 6 - 7 | 729 | 89 | |
| GenBank mRNA | BC065611.1 | - | 6 - 7 | 725 | 89 | |
| U55177.1 | - | 6 - 7 | 729 | 89 | ||
More Information
Set Membership: |
|

cahz
Genomic Map