Assay Details
| Gene Symbol: ctnnbip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131594.1 | NP_571669.1 | 2 - 3 | 218 | 74 | |
| GenBank mRNA | AB046866.1 | - | 2 - 3 | 218 | 74 | |
| BC054693.1 | - | 2 - 3 | 187 | 74 | ||
More Information
Set Membership: |
|

ctnnbip1
Genomic Map